Learn More
4-(Methylamino)benzoic acid, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1242614
Description
4-(Methylamino)benzoic acid important for the preparation of other pharmaceutical products, dyes, flavors, and preservatives. .For the preparation of medicines and other fine chemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-(Methylamino)benzoic acid important for the preparation of other pharmaceutical products, dyes, flavors, and preservatives. .For the preparation of medicines and other fine chemicals.
Solubility
Slightly soluble. Soluble in alcohol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
4- (Methylamino)benzoic acid | |
158°C to 160°C | |
MFCD00002535 | |
2082805 | |
Slightly soluble. Soluble in alcohol. | |
CNC1=CC=C(C=C1)C(=O)O | |
151.165 | |
CHEBI:7308 | |
97% |
10541-83-0 | |
C8H9NO2 | |
25 g | |
4-methylamino benzoic acid, benzoic acid, 4-methylamino, p-methylaminobenzoic acid, n-methyl-4-aminobenzoic acid, 4-n-methylamino benzoic acid, n-methyl-4-aminobenzoate, benzoic acid, p-methylamino, 4-methylamino-benzoic acid, p-methylamino benzoic acid, p-n-methylamino benzoic acid | |
ZVIDMSBTYRSMAR-UHFFFAOYSA-N | |
4-(methylamino)benzoic acid | |
66345 | |
151.17 |
Chemical Identifiers
10541-83-0 | |
151.165 | |
ZVIDMSBTYRSMAR-UHFFFAOYSA-N | |
66345 | |
4-(methylamino)benzoic acid |
C8H9NO2 | |
MFCD00002535 | |
4-methylamino benzoic acid, benzoic acid, 4-methylamino, p-methylaminobenzoic acid, n-methyl-4-aminobenzoic acid, 4-n-methylamino benzoic acid, n-methyl-4-aminobenzoate, benzoic acid, p-methylamino, 4-methylamino-benzoic acid, p-methylamino benzoic acid, p-n-methylamino benzoic acid | |
CHEBI:7308 | |
CNC1=CC=C(C=C1)C(=O)O |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 234-121-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only