missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4′-Methylacetophenone 95.0+%, TCI America™

Manufacturer:  Tci America M0135500ML

 View more versions of this product

Catalog No. M0135500ML



Specifications

Specifications

4′-Methylacetophenone
Yellow
122-00-9
MFCD00008751
GNKZMNRKLCTJAY-UHFFFAOYSA-N
1-(4-methylphenyl)ethanone
8500
Clear Liquid at 20°C
226°C
500mL
C9H10O
1-4-methylphenyl ethanone, 1-p-tolyl ethanone, 1-p-tolylethanone, 4'-methylacetophenone, 4-acetyltoluene, 4-methylacetophenone, melilotal, methyl p-tolyl ketone, p-acetotoluene, p-methylacetophenone
CC1=CC=C(C=C1)C(=O)C
134.178
134.18
≥95.0% (GC)

Chemical Identifiers

122-00-9
134.178
GNKZMNRKLCTJAY-UHFFFAOYSA-N
8500
CC1=CC=C(C=C1)C(=O)C
C9H10O
MFCD00008751
1-4-methylphenyl ethanone, 1-p-tolyl ethanone, 1-p-tolylethanone, 4'-methylacetophenone, 4-acetyltoluene, 4-methylacetophenone, melilotal, methyl p-tolyl ketone, p-acetotoluene, p-methylacetophenone
1-(4-methylphenyl)ethanone
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1239

RTECSNumber : AM9463000

TSCA : Yes

Documents