Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-Methylacetanilide, 98+%

$27.43 - $27.43

Chemical Identifiers

Chemical Name or Material 4'-Methylacetanilide
CAS 103-89-9
Molecular Formula C9H11NO
Formula Weight 149.19
MDL Number MFCD00008677
Beilstein 607036
Merck Index 14,74
Synonym p-Acetotoluidide; 4-Acetamidotoluene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1372114

  SDS
alfa aesar™
A1372114
25g Each for $27.43
Description & Specifications

Specifications

Formula Weight 149.19
Quantity 25g
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Beilstein 607036
Merck Index 14,74
TSCA TSCA
CAS 103-89-9
RTECS Number AN2930000
Synonym p-Acetotoluidide; 4-Acetamidotoluene
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay ≥98%
Chemical Name or Material 4'-Methylacetanilide
Molecular Formula C9H11NO
Boiling Point 306° to 308°C
Melting Point 147° to 150°C
MDL Number MFCD00008677
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.