missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methyl-L-phenylalanine, TCI America™

$131.39 - $455.28

Chemical Identifiers

CAS 1991-87-3
Molecular Formula C10H13NO2
Molecular Weight (g/mol) 179.219
MDL Number MFCD00038575
InChI Key DQLHSFUMICQIMB-VIFPVBQESA-N
Synonym 4-methyl-l-phenylalanine, l-4-methylphenylalanine, 4-methylphenylalanine, h-phe 4-me-oh, 2s-2-amino-3-4-methylphenyl propanoic acid, phenylalanine, 4-methyl, s-2-amino-3-p-tolyl propanoic acid, l-4-me-phe-oh, l-4-methylphe, 4-methyl-l-phe-oh
PubChem CID 151513
IUPAC Name (2S)-2-amino-3-(4-methylphenyl)propanoic acid
SMILES CC1=CC=C(C=C1)CC(C(=O)O)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M27435G View Documents TCI America
M27435G
5g Each for $455.28
 
M27431G View Documents TCI America
M27431G
1g Each for $131.39
 
Specifications

Chemical Identifiers

1991-87-3
179.219
DQLHSFUMICQIMB-VIFPVBQESA-N
151513
CC1=CC=C(C=C1)CC(C(=O)O)N
C10H13NO2
MFCD00038575
4-methyl-l-phenylalanine, l-4-methylphenylalanine, 4-methylphenylalanine, h-phe 4-me-oh, 2s-2-amino-3-4-methylphenyl propanoic acid, phenylalanine, 4-methyl, s-2-amino-3-p-tolyl propanoic acid, l-4-me-phe-oh, l-4-methylphe, 4-methyl-l-phe-oh
(2S)-2-amino-3-(4-methylphenyl)propanoic acid

Specifications

White-Yellow
5g
C10H13NO2
4-methyl-l-phenylalanine, l-4-methylphenylalanine, 4-methylphenylalanine, h-phe 4-me-oh, 2s-2-amino-3-4-methylphenyl propanoic acid, phenylalanine, 4-methyl, s-2-amino-3-p-tolyl propanoic acid, l-4-me-phe-oh, l-4-methylphe, 4-methyl-l-phe-oh
CC1=CC=C(C=C1)CC(C(=O)O)N
179.219
179.22
≥98.0% (HPLC,T)
271°C
1991-87-3
MFCD00038575
DQLHSFUMICQIMB-VIFPVBQESA-N
(2S)-2-amino-3-(4-methylphenyl)propanoic acid
151513
Crystal-Powder at 20°C
4-Methyl-L-phenylalanine
Safety and Handling

Safety and Handling

TSCA : No