Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methyl-L-phenylalanine, 98%

$41.88 - $41.88

Chemical Identifiers

Chemical Name or Material 4-Methyl-L-phenylalanine
CAS 1991-87-3
Molecular Formula C10H13NO2
Formula Weight 179.22
MDL Number MFCD00038575
Synonym H-Phe(4-Me)-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH26905MD

  SDS
alfa aesar™
H26905MD
250mg Each for $41.88
Description & Specifications

Specifications

Formula Weight 179.22
Quantity 250mg
CAS 1991-87-3
Synonym H-Phe(4-Me)-OH
Chemical Name or Material 4-Methyl-L-phenylalanine
Molecular Formula C10H13NO2
Assay 98%
MDL Number MFCD00038575

Applications
Used in absorption liquids on a technical scale for removing acid components such as carbon dioxide and hydrogen sulfide from gases. 4-Methyl-L-phenylalanine are used in buffers to maintain the pH of aqueous liquids at a constant level.

Solubility
Sparingly soluble (0.014 g/L) (25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.