missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Methyl-D-phenylalanine, 98%, Thermo Scientific™
$366.45 - $366.45
Chemical Identifiers
CAS | 49759-61-7 |
---|---|
Molecular Formula | C10H13NO2 |
Molecular Weight (g/mol) | 179.22 |
MDL Number | MFCD00152137 |
InChI Key | DQLHSFUMICQIMB-UHFFFAOYNA-N |
Synonym | 4-methyl-d-phenylalanine, h-d-phe 4-me-oh, r-2-amino-3-p-tolyl propanoic acid, d-4-methylphenylalanine, d-4-me-phe-oh, 2r-2-amino-3-4-methylphenyl propanoic acid, d-phenylalanine, 4-methyl, h-p-me-phe-oh, 4-methylphenyl-d-alanine, d-4-methylphe |
PubChem CID | 7015862 |
IUPAC Name | (2R)-2-amino-3-(4-methylphenyl)propanoic acid |
SMILES | CC1=CC=C(CC(N)C(O)=O)C=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH2751803
|
Thermo Scientific Chemicals
H2751803 |
1 g |
Each for $366.45
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
49759-61-7 | |
179.22 | |
DQLHSFUMICQIMB-UHFFFAOYNA-N | |
7015862 | |
CC1=CC=C(CC(N)C(O)=O)C=C1 |
C10H13NO2 | |
MFCD00152137 | |
4-methyl-d-phenylalanine, h-d-phe 4-me-oh, r-2-amino-3-p-tolyl propanoic acid, d-4-methylphenylalanine, d-4-me-phe-oh, 2r-2-amino-3-4-methylphenyl propanoic acid, d-phenylalanine, 4-methyl, h-p-me-phe-oh, 4-methylphenyl-d-alanine, d-4-methylphe | |
(2R)-2-amino-3-(4-methylphenyl)propanoic acid |
Specifications
49759-61-7 | |
MFCD00152137 | |
DQLHSFUMICQIMB-UHFFFAOYNA-N | |
(2R)-2-amino-3-(4-methylphenyl)propanoic acid | |
7015862 | |
98% | |
4-Methyl-D-phenylalanine |
C10H13NO2 | |
4-methyl-d-phenylalanine, h-d-phe 4-me-oh, r-2-amino-3-p-tolyl propanoic acid, d-4-methylphenylalanine, d-4-me-phe-oh, 2r-2-amino-3-4-methylphenyl propanoic acid, d-phenylalanine, 4-methyl, h-p-me-phe-oh, 4-methylphenyl-d-alanine, d-4-methylphe | |
CC1=CC=C(CC(N)C(O)=O)C=C1 | |
179.22 | |
179.22 | |
1 g |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures