missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methyl-7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,4-dihydro-2H-pyrido[3,2-b][1,4, Maybridge

Chemical Identifiers

CAS 910037-15-9
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 90%
Molecular Formula C14H21BN2O3
Formula Weight 276.14
MDL Number MFCD09065020
Chemical Name or Material 4-Methyl-7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,4-dihydro-2H-pyrido[3,2-b][1,4]oxazine
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC62039CB SDS Fisher Scientific Company
CC62039CB
Amber Glass Bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC62039DA SDS Fisher Scientific Company
CC62039DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

910037-15-9
100.0
C14H21BN2O3
MFCD09065020
97%
96.5
90%
276.14
4-Methyl-7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,4-dihydro-2H-pyrido[3,2-b][1,4]oxazine

Specifications

276.14
100.0
C14H21BN2O3
MFCD09065020
910037-15-9
97%
250mg
Amber Glass Bottle
96.5
90%
4-Methyl-7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,4-dihydro-2H-pyrido[3,2-b][1,4]oxazine

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning