missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Methyl-7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,4-dihydro-2H-1,4-benzoxazine, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC13539DA
Specifications
4-Methyl-7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,4-dihydro-2H-1,4-benzoxazine | |
97% | |
C15H22BNO3 | |
1 g | |
QRAOZQGIUIDZQZ-UHFFFAOYSA-N | |
4-methyl-7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,3-dihydro-1,4-benzoxazine | |
2795301 |
519054-54-7 | |
Amber Glass Bottle | |
MFCD04115377 | |
4-methyl-7-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3,4-dihydro-2h-1,4-benzoxazine, 4-methyl-7-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3,4-dihydro-2h-benzo b 1,4 oxazine, 4-methyl-3,4-dihydro-2h-1,4-benzoxazin-7-ylboronic acid pinacol ester, 4-methyl-7-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2,3-dihydro-1,4-benzoxazine, 4-methyl-7-tetramethyl-1,3,2-dioxaborolan-2-yl-2,3-dihydro-1,4-benzoxazine, 4-methyl-7-4,4,5,5-tetramethyl-1,3,2-dioxaborolan, 4-methyl-7-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3,4-dihydro-2h-1,4-benzoxazine, 4-methyl-7-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3,4-dihydro-2h-benzo 1,4 oxazine, 4-methyl-7-4,4,5,5-tetramethyl-1,3,2,-dioxaborolan-2-yl-3,4-dihydro-2h-1,4-benzoxazine, 4-methyl-7-4,4,5,5-tetramethyl-1,3,2-d ioxaborolan-2-yl-3,4-dihydro-2h-benzo b 1,4 oxazine | |
CN1CCOC2=C1C=CC(=C2)B1OC(C)(C)C(C)(C)O1 | |
275.16 | |
275.15 |
Chemical Identifiers
519054-54-7 | |
275.16 | |
QRAOZQGIUIDZQZ-UHFFFAOYSA-N | |
2795301 | |
CN1CCOC2=C1C=CC(=C2)B1OC(C)(C)C(C)(C)O1 |
C15H22BNO3 | |
MFCD04115377 | |
4-methyl-7-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3,4-dihydro-2h-1,4-benzoxazine, 4-methyl-7-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3,4-dihydro-2h-benzo b 1,4 oxazine, 4-methyl-3,4-dihydro-2h-1,4-benzoxazin-7-ylboronic acid pinacol ester, 4-methyl-7-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2,3-dihydro-1,4-benzoxazine, 4-methyl-7-tetramethyl-1,3,2-dioxaborolan-2-yl-2,3-dihydro-1,4-benzoxazine, 4-methyl-7-4,4,5,5-tetramethyl-1,3,2-dioxaborolan, 4-methyl-7-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3,4-dihydro-2h-1,4-benzoxazine, 4-methyl-7-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3,4-dihydro-2h-benzo 1,4 oxazine, 4-methyl-7-4,4,5,5-tetramethyl-1,3,2,-dioxaborolan-2-yl-3,4-dihydro-2h-1,4-benzoxazine, 4-methyl-7-4,4,5,5-tetramethyl-1,3,2-d ioxaborolan-2-yl-3,4-dihydro-2h-benzo b 1,4 oxazine | |
4-methyl-7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,3-dihydro-1,4-benzoxazine |