Please login to your online account to display your discounted pricing

4-Methyl-5-thiazolylethyl acetate, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Methyl-5-thiazolylethyl acetate
Name Note 99%
CAS 656-53-1
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C8H11NO2S
Formula Weight 185.25
MDL Number MFCD00005338
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC265400050

  SDS
thermo fisher scientific
265400050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Liquid
Formula Weight 185.25
Quantity 5g
Color Clear brown
EINECS Number 211-515-7
TSCA TSCA
CAS Max % 100.0
CAS Min % 98.5
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 4-Methyl-5-thiazolylethyl acetate
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 99%
Refractive Index 1.5090 to 1.5110 (20°C, 589nm)
Assay Percent Range 99%
Molecular Formula C8H11NO2S
MDL Number MFCD00005338
CAS 656-53-1
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.