Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methyl-5-thiazolemethanol, 97%

Chemical Identifiers

Chemical Name or Material 4-Methyl-5-thiazolemethanol
CAS 1977-06-6
Formula Weight 129.18
Synonym (4-Methyl-1,3-thiazol-5-yl)methanol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6190003

 
alfa aesar™
H6190003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH61900MD

 
alfa aesar™
H61900MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 129.18
Chemical Name or Material 4-Methyl-5-thiazolemethanol
Physical Form Solid
Assay 97%
CAS 1977-06-6
Synonym (4-Methyl-1,3-thiazol-5-yl)methanol
Quantity 1g