Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methyl-5-thiazoleethanol, 98%

$39.32 - $105.03

Chemical Identifiers

Chemical Name or Material 4-Methyl-5-thiazoleethanol
CAS 137-00-8
Molecular Formula C6H9NOS
Formula Weight 143.21
MDL Number MFCD00005339
Beilstein 114249
Merck Index 14,6126
Synonym 5-(2-Hydroxyethyl)-4-methylthiazole; Sulfurol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1356714

  SDS
alfa aesar™
A1356714
25g Each for $39.32

AAA1356722

  SDS
alfa aesar™
A1356722
100g Each for $105.03
Description & Specifications

Specifications

Formula Weight 143.21
Chemical Name or Material 4-Methyl-5-thiazoleethanol
TSCA TSCA
Assay 98%
Molecular Formula C6H9NOS
MDL Number MFCD00005339
CAS 137-00-8
Beilstein 114249
Merck Index 14,6126
Synonym 5-(2-Hydroxyethyl)-4-methylthiazole; Sulfurol
Health Hazard 1 Warning
Boiling Point 135° to 137°C (7mmHg)
Quantity 25g
Flash Point 142°C (287°F)
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.