Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methyl-5-nonanone, 99+%

$86.66 - $995.92

Chemical Identifiers

Chemical Name or Material 4-Methyl-5-nonanone
CAS 35900-26-6
Molecular Formula C10H20O
Formula Weight 156.27
MDL Number MFCD00048807
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2321906

  SDS
alfa aesar™
B2321906
5g Each for $86.66

AAB2321914

  SDS
alfa aesar™
B2321914
25g Each for $317.09

AAB2321922

  SDS
alfa aesar™
B2321922
100g Each for $995.92
Description & Specifications

Specifications

Formula Weight 156.27
Chemical Name or Material 4-Methyl-5-nonanone
TSCA TSCA
Assay ≥99%
Molecular Formula C10H20O
MDL Number MFCD00048807
CAS 35900-26-6
Boiling Point 189° to 190°C
Quantity 5g