Please login to your online account to display your discounted pricing

4-Methyl-5-imidazolemethanol hydrochloride, 98%, ACROS Organics™


Quantity 25g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  210880250

 View more versions of this product

Catalog No. AC210880250

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 148.59
Quantity 25g
Color Very slightly beige
Beilstein 23, I, 105
EINECS Number 254-021-7
Melting Point 231° to 235°C
CAS Max % 100.0
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS Min % 97.5
Packaging Plastic bottle
Infrared Spectrum Authentic
Health Hazard 1 HYGROSCOPIC
Name Note 98%
Molecular Formula C5H8N2O·HCl
MDL Number MFCD00040986
CAS 38585-62-5
Assay Percent Range 98%
Chemical Name or Material 4-Methyl-5-imidazolemethanol hydrochloride

Specifications

Chemical Name or Material 4-Methyl-5-imidazolemethanol hydrochloride
Name Note 98%
CAS 38585-62-5
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C5H8N2O·HCl
Formula Weight 148.59
MDL Number MFCD00040986
Accessories

HYGROSCOPIC

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents