missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methyl-5-imidazolemethanol hydrochloride, 98%, ACROS Organics™

$54.08 - $54.08

Chemical Identifiers

CAS 38585-62-5
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C5H8N2O·HCl
Formula Weight 148.59
MDL Number MFCD00040986
Chemical Name or Material 4-Methyl-5-imidazolemethanol hydrochloride, 98%
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC210880250 SDS Thermo Fisher Scientific
210880250
Plastic bottle 25g Each for $54.08
Specifications Description & Specifications

Chemical Identifiers

38585-62-5
100.0
C5H8N2O·HCl
MFCD00040986
≥97.5%
97.5
98%
148.59
4-Methyl-5-imidazolemethanol hydrochloride, 98%

Specifications

38585-62-5
100.0
Plastic bottle
98%
148.59
23, I, 105
231° to 235°C
4-Methyl-5-imidazolemethanol hydrochloride, 98%
97.5
Authentic
Crystalline Powder
C5H8N2O·HCl
MFCD00040986
Very slightly beige
25g
≥97.5%

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

HYGROSCOPIC

EINECSNumber : 254-021-7