Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methyl-5-(2-pyrazinyl)-1,2-dithiole-3-thione

Chemical Identifiers

Chemical Name or Material 4-Methyl-5-(2-pyrazinyl)-1,2-dithiole-3-thione
CAS 64224-21-1
Formula Weight 226.34
MDL Number MFCD00868499
Synonym Oltipraz
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH35120MC

  SDS
alfa aesar™
H35120MC
100mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH35120ME

  SDS
alfa aesar™
H35120ME
500mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 226.34
Chemical Name or Material 4-Methyl-5-(2-pyrazinyl)-1,2-dithiole-3-thione
Physical Form Solid
MDL Number MFCD00868499
CAS 64224-21-1
Synonym Oltipraz
Color Orange-Red
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Storage Note 1 2° to 8°C
RTECS Number JP1293000
Sensitivity Light Sensitive
Quantity 100mg
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C