missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methyl-5-(2-pyrazinyl)-1,2-dithiole-3-thione, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H35120MC

 View more versions of this product

Catalog No. AAH35120MC



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Methyl-5-(2-pyrazinyl)-1,2-dithiole-3-thione
Orange-Red
MFCD00868499
Light sensitive
CKNAQFVBEHDJQV-UHFFFAOYSA-N
4-methyl-5-pyrazin-2-yldithiole-3-thione
47318
226.34
64224-21-1
C8H6N2S3
100 mg
oltipraz, oltiprazum, 4-methyl-5-pyrazin-2-yl-3h-1,2-dithiole-3-thione, ccris 4048, oltipraz inn, 4-methyl-5-pyrazinyl-3h-1,2-dithiole-3-thione, oltiprazum inn-latin, unii-6n510jul1y, 5-2-pyrazinyl-4-methyl-1,2-dithiole-3-thione, 3h-1,2-dithiole-3-thione, 4-methyl-5-pyrazinyl
CC1=C(SSC1=S)C1=CN=CC=N1
226.33
CHEBI:77319
Solid

Chemical Identifiers

64224-21-1
226.33
CKNAQFVBEHDJQV-UHFFFAOYSA-N
47318
4-methyl-5-pyrazin-2-yldithiole-3-thione
C8H6N2S3
MFCD00868499
oltipraz, oltiprazum, 4-methyl-5-pyrazin-2-yl-3h-1,2-dithiole-3-thione, ccris 4048, oltipraz inn, 4-methyl-5-pyrazinyl-3h-1,2-dithiole-3-thione, oltiprazum inn-latin, unii-6n510jul1y, 5-2-pyrazinyl-4-methyl-1,2-dithiole-3-thione, 3h-1,2-dithiole-3-thione, 4-methyl-5-pyrazinyl
CHEBI:77319
CC1=C(SSC1=S)C1=CN=CC=N1
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 264-736-6

RTECSNumber : JP1293000

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only