missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methyl-5-(2-pyrazinyl)-1,2-dithiole-3-thione

Manufacturer:  Alfa Aesar™ H35120ME

 View more versions of this product

Catalog No. AAH35120ME


 

Specifications

Specifications

4-Methyl-5-(2-pyrazinyl)-1,2-dithiole-3-thione
500mg
C8H6N2S3
Oltipraz
CKNAQFVBEHDJQV-UHFFFAOYSA-N
4-methyl-5-pyrazin-2-yldithiole-3-thione
47318
226.34
Light Sensitive
64224-21-1
MFCD00868499
Orange-Red
CC1=C(SSC1=S)C2=NC=CN=C2
226.33
CHEBI:77319
Solid

Chemical Identifiers

64224-21-1
226.33
CKNAQFVBEHDJQV-UHFFFAOYSA-N
47318
C8H6N2S3
MFCD00868499
Oltipraz
CHEBI:77319
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : JP1293000

storageNote1 : 2° to 8°C

SDS Documents