missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methyl-4-nitrovaleraldehyde, TCI America™

$177.22 - $603.48

Chemical Identifiers

CAS 57620-49-2
Molecular Formula C6H11NO3
Molecular Weight (g/mol) 145.158
InChI Key YDVAFDALOGXWTO-UHFFFAOYSA-N
Synonym 4-Methyl-4-nitropentanal
PubChem CID 12264009
IUPAC Name 4-methyl-4-nitropentanal
SMILES CC(C)(CCC=O)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M182525G View Documents TCI America
M182525G
25g Each for $603.48
 
M18255G View Documents TCI America
M18255G
5g Each for $177.22
 
Specifications

Chemical Identifiers

57620-49-2
145.158
4-Methyl-4-nitropentanal
4-methyl-4-nitropentanal
C6H11NO3
YDVAFDALOGXWTO-UHFFFAOYSA-N
12264009
CC(C)(CCC=O)[N+](=O)[O-]

Specifications

90°C
25g
C6H11NO3
YDVAFDALOGXWTO-UHFFFAOYSA-N
4-methyl-4-nitropentanal
12264009
Clear Liquid at 20°C
4-Methyl-4-nitrovaleraldehyde
Undesignated
57620-49-2
4-Methyl-4-nitropentanal
CC(C)(CCC=O)[N+](=O)[O-]
145.158
145.16
≥90.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No