Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methyl-4-methylthio-2-pentanone, 99%

$324.78 - $324.78

Chemical Identifiers

Chemical Name or Material 4-Methyl-4-methylthio-2-pentanone
CAS 23550-40-5
Molecular Formula C6H8OS
Formula Weight 146.25
MDL Number MFCD00049253
Synonym 4-Methyl-4-(methylmercapto)pentan-2-one
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2205822

 
alfa aesar™
B2205822
100g Each for $324.78
Description & Specifications

Specifications

Formula Weight 146.25
Quantity 100g
Chemical Name or Material 4-Methyl-4-methylthio-2-pentanone
TSCA TSCA
Assay 99%
Molecular Formula C6H8OS
MDL Number MFCD00049253
CAS 23550-40-5
Synonym 4-Methyl-4-(methylmercapto)pentan-2-one
Health Hazard 1 Warning
Boiling Point 80° to 82°C (10mmHg)
Flash Point 72°C (162°F)
Health Hazard 2 GHS H Statement
H227
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H227
Combustible liquid.