Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methyl-4-hepten-3-ol, 97%

Chemical Identifiers

Chemical Name or Material 4-Methyl-4-hepten-3-ol
CAS 81280-12-8
Molecular Formula C8H16O
Formula Weight 128.22
MDL Number MFCD00046670
UN Number UN1987
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2065803

  SDS
alfa aesar™
B2065803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2065806

  SDS
alfa aesar™
B2065806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2065814

  SDS
alfa aesar™
B2065814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 128.22
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Chemical Name or Material 4-Methyl-4-hepten-3-ol
Flash Point 59°C (138°F)
Molecular Formula C8H16O
Quantity 1g
Health Hazard 2 GHS H Statement
H226
Flammable liquid and vapour.
CAS 81280-12-8
Assay 97%
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
UN Number UN1987
MDL Number MFCD00046670
Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226
Flammable liquid and vapour.