Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methyl-3-(trifluoromethyl)phenol, 97%

Chemical Identifiers

Chemical Name or Material 4-Methyl-3-(trifluoromethyl)phenol
Molecular Formula C8H7F3O
Formula Weight 176.14
MDL Number MFCD09832294
UN Number UN3261
Synonym 5-Hydroxy-2-methylbenzotrifluoride; 3-Trifluoromethyl-p-cresol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3205103

  SDS
alfa aesar™
H3205103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH32051MD

  SDS
alfa aesar™
H32051MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 176.14
Chemical Name or Material 4-Methyl-3-(trifluoromethyl)phenol
Assay 97%
Molecular Formula C8H7F3O
MDL Number MFCD09832294
UN Number UN3261
Synonym 5-Hydroxy-2-methylbenzotrifluoride; 3-Trifluoromethyl-p-cresol
Health Hazard 1 Danger
Melting Point 45° to 48°C
Quantity 1g
Health Hazard 2 GHS H Statement
H314-H318-H302-H312
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318-H302-H312
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.