Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methyl-3-(trifluoromethyl)-DL-phenylalanine, 97%

Chemical Identifiers

Chemical Name or Material 4-Methyl-3-(trifluoromethyl)-DL-phenylalanine
Molecular Formula C11H12F3NO2
Formula Weight 247.22
MDL Number MFCD09832296
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3219703

 
alfa aesar™
H3219703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH32197MD

 
alfa aesar™
H32197MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 247.22
Chemical Name or Material 4-Methyl-3-(trifluoromethyl)-DL-phenylalanine
Assay 97%
Molecular Formula C11H12F3NO2
MDL Number MFCD09832296
Melting Point ≈237°C
Quantity 1g