Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methyl-3-(trifluoromethyl)benzoic acid, 97+%

$110.14 - $394.77

Chemical Identifiers

Chemical Name or Material 4-Methyl-3-(trifluoromethyl)benzoic acid
CAS 261952-01-6
Molecular Formula C9H7F3O2
Formula Weight 204.15
MDL Number MFCD01631592
Synonym 3-Trifluoromethyl-p-toluic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2424603

  SDS
alfa aesar™
B2424603
1g Each for $110.14

AAB2424606

  SDS
alfa aesar™
B2424606
5g Each for $394.77
Description & Specifications

Specifications

Formula Weight 204.15
Melting Point 175° to 178°C
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1g
Assay ≥97%
Chemical Name or Material 4-Methyl-3-(trifluoromethyl)benzoic acid
Health Hazard 1 Warning
CAS 261952-01-6
MDL Number MFCD01631592
Molecular Formula C9H7F3O2
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Synonym 3-Trifluoromethyl-p-toluic acid
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.