Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methyl-3-(trifluoromethyl)benzaldehyde, 97%

$190.86 - $575.69

Chemical Identifiers

Chemical Name or Material 4-Methyl-3-(trifluoromethyl)benzaldehyde
CAS 93249-45-7
Molecular Formula C9H7F3O
Formula Weight 188.15
MDL Number MFCD04116033
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2676203

 
alfa aesar™
H2676203
1g Each for $190.86

AAH2676206

 
alfa aesar™
H2676206
5g Each for $575.69
Description & Specifications

Specifications

Formula Weight 188.15
Chemical Name or Material 4-Methyl-3-(trifluoromethyl)benzaldehyde
Assay 97%
Molecular Formula C9H7F3O
MDL Number MFCD04116033
CAS 93249-45-7
Health Hazard 1 Warning
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
4-Methyl-3-(trifluoromethyl)benzaldehyde is used as a building block in synthetic chemistry.

Notes
Air sensitive. Incompatible with strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.