Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methyl-3-(trifluoromethyl)aniline, 98+%

Chemical Identifiers

Chemical Name or Material 4-Methyl-3-(trifluoromethyl)aniline
CAS 65934-74-9
Molecular Formula C8H8F3N
Formula Weight 175.16
MDL Number MFCD01631582
UN Number UN2810
Beilstein 6776230
Synonym 5-Amino-2-methylbenzotrifluoride; 3-Trifluoromethyl-p-toluidine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2339603

  SDS
alfa aesar™
B2339603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2339606

  SDS
alfa aesar™
B2339606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2339614

  SDS
alfa aesar™
B2339614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 175.16
CAS 65934-74-9
UN Number UN2810
Flash Point 102°C (216°F)
Molecular Formula C8H8F3N
Quantity 1g
Chemical Name or Material 4-Methyl-3-(trifluoromethyl)aniline
Health Hazard 2 GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Beilstein 6776230
Health Hazard 1 Danger
MDL Number MFCD01631582
Synonym 5-Amino-2-methylbenzotrifluoride; 3-Trifluoromethyl-p-toluidine
Assay ≥98%
Boiling Point 204°C
Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.