Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methyl-3-thiosemicarbazide, 97%

Chemical Identifiers

Chemical Name or Material 4-Methyl-3-thiosemicarbazide
CAS 6610-29-3
Molecular Formula C2H7N3S
Formula Weight 105.16
MDL Number MFCD00007617
UN Number UN2811
Beilstein 605575
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2113809

  SDS
alfa aesar™
B2113809
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2113818

  SDS
alfa aesar™
B2113818
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2113830

  SDS
alfa aesar™
B2113830
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 105.16
Health Hazard 3 GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 4-Methyl-3-thiosemicarbazide
TSCA TSCA
Assay 97%
Molecular Formula C2H7N3S
MDL Number MFCD00007617
CAS 6610-29-3
UN Number UN2811
Beilstein 605575
Health Hazard 1 Danger
Melting Point 135° to 138°C
Quantity 10g
Health Hazard 2 GHS H Statement
H300
Fatal if swallowed.
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
RTECS Number MV1407370

Applications
4-Methyl-3-thiosemicarbazide is used in the analysis of electronic features of sulfur-using high-resolution, low-temperature X-ray diffraction data sets. Further, it acts as an intermediate in organic synthesis.

Solubility
Soluble in alcohol. Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents, strong acids and bases.

Safety and Handling

Danger

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H300
Fatal if swallowed.