missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methyl-3-nitrothiobenzamide, 97%

Chemical Identifiers

CAS 1233513-42-2
Molecular Formula C8H8N2O2S
Molecular Weight (g/mol) 196.224
MDL Number MFCD09025715
InChI Key PGQMEJDSPAENNP-UHFFFAOYSA-N
Synonym 4-methyl-3-nitrothiobenzamide
PubChem CID 45040726
IUPAC Name 4-methyl-3-nitrobenzenecarbothioamide
SMILES CC1=C(C=C(C=C1)C(=S)N)[N+](=O)[O-]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH54709MD View Documents Alfa Aesar™
H54709MD
250mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH5470903 View Documents Alfa Aesar™
H5470903
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH5470906 View Documents Alfa Aesar™
H5470906
5g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

1233513-42-2
196.224
PGQMEJDSPAENNP-UHFFFAOYSA-N
45040726
CC1=C(C=C(C=C1)C(=S)N)[N+](=O)[O-]
C8H8N2O2S
MFCD09025715
4-methyl-3-nitrothiobenzamide
4-methyl-3-nitrobenzenecarbothioamide

Specifications

250mg
C8H8N2O2S
4-methyl-3-nitrothiobenzamide
CC1=C(C=C(C=C1)C(=S)N)[N+](=O)[O-]
196.224
196.22
4-Methyl-3-nitrothiobenzamide
1233513-42-2
MFCD09025715
PGQMEJDSPAENNP-UHFFFAOYSA-N
4-methyl-3-nitrobenzenecarbothioamide
45040726
97%
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger