missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methyl-3-nitropyridine, TCI America™

$97.78 - $336.13

Chemical Identifiers

CAS 5832-44-0
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.126
MDL Number MFCD00051829
InChI Key JLNRJMGYBKMDGI-UHFFFAOYSA-N
Synonym 3-nitro-4-picoline, 4-methyl-3-nitro-pyridine, 3-nitro-4-methylpyridine, pyridine, 4-methyl-3-nitro, zlchem 957, pubchem6644, 4-methyl-3-nitro pyridine, acmc-1b13v, ksc490s2l, tpc-py011
PubChem CID 521828
IUPAC Name 4-methyl-3-nitropyridine
SMILES CC1=C(C=NC=C1)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M23535G View Documents TCI America
M23535G
5g Each for $336.13
 
M23531G View Documents TCI America
M23531G
1g Each for $97.78
 
Specifications

Chemical Identifiers

5832-44-0
138.126
JLNRJMGYBKMDGI-UHFFFAOYSA-N
521828
CC1=C(C=NC=C1)[N+](=O)[O-]
C6H6N2O2
MFCD00051829
3-nitro-4-picoline, 4-methyl-3-nitro-pyridine, 3-nitro-4-methylpyridine, pyridine, 4-methyl-3-nitro, zlchem 957, pubchem6644, 4-methyl-3-nitro pyridine, acmc-1b13v, ksc490s2l, tpc-py011
4-methyl-3-nitropyridine

Specifications

110°C
5g
C6H6N2O2
3-nitro-4-picoline, 4-methyl-3-nitro-pyridine, 3-nitro-4-methylpyridine, pyridine, 4-methyl-3-nitro, zlchem 957, pubchem6644, 4-methyl-3-nitro pyridine, acmc-1b13v, ksc490s2l, tpc-py011
CC1=C(C=NC=C1)[N+](=O)[O-]
138.126
138.13
≥97.0% (GC)
Yellow
5832-44-0
MFCD00051829
JLNRJMGYBKMDGI-UHFFFAOYSA-N
4-methyl-3-nitropyridine
521828
Crystal-Lump at 20°C
4-Methyl-3-nitropyridine
Safety and Handling

Safety and Handling

TSCA : No