Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methyl-3-nitropyridine, 98%

Chemical Identifiers

Chemical Name or Material 4-Methyl-3-nitropyridine
CAS 5832-44-0
Molecular Formula C6H6N2O2
Formula Weight 138.13
MDL Number MFCD00051829
UN Number UN2810
Beilstein 118796
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0739803

 
alfa aesar™
L0739803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0739806

 
alfa aesar™
L0739806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 138.13
Chemical Name or Material 4-Methyl-3-nitropyridine
CAS 5832-44-0
Assay 98%
Molecular Formula C6H6N2O2
MDL Number MFCD00051829
UN Number UN2810
Beilstein 118796
Health Hazard 1 Danger
Melting Point 18° to 22°C
Boiling Point 238°C
Quantity 1g
Flash Point 108°C (226°F)
Health Hazard 2 GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

2° to 8°C