Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methyl-3-nitrophenyl isocyanate, 96%

Chemical Identifiers

Chemical Name or Material 4-Methyl-3-nitrophenyl isocyanate
CAS 13471-69-7
Molecular Formula C8H6N2O3
Formula Weight 178.15
MDL Number MFCD00014698
UN Number UN2811
Beilstein 2724128
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1266903

  SDS
alfa aesar™
L1266903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1266906

  SDS
alfa aesar™
L1266906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 178.15
Chemical Name or Material 4-Methyl-3-nitrophenyl isocyanate
TSCA TSCA
Assay 96%
MDL Number MFCD00014698
CAS 13471-69-7
Beilstein 2724128
Health Hazard 1 Danger
Boiling Point 113° to 118°C (3mmHg)
Quantity 1g
Flash Point >110°C (230°F)
Health Hazard 2 GHS H Statement
H331-H334-H302-H312-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Molecular Formula C8H6N2O3
UN Number UN2811
Melting Point 48° to 54°C
Odor Irritating
Safety and Handling

Danger

GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H334-H302-H312-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C