missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methyl-3-nitrophenol, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 04428518

 View more versions of this product

Catalog No. AA4428518



Description

Description

It is used as a chemical, pharmaceutical and OLED intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Methyl-3-nitrophenol
77°C to 81°C
C7H7NO3
50 g
3-nitro-4-methylphenol, 3-nitro-p-cresol, 4-methyl-3-nitro-phenol, 4-hydroxy-2-nitrotoluene, 2-nitro-4-hydroxytoluene, phenol, 4-methyl-3-nitro, p-cresol, 3-nitro, unii-4b422hud6h, 4-methyl-5-nitrophenol
BQEXDUKMTVYBRK-UHFFFAOYSA-N
4-methyl-3-nitrophenol
16271
98%
2042-14-0
Phenol-like
MFCD00007244
UN2446
Slightly soluble in water.
CC1=C(C=C(C=C1)O)[N+](=O)[O-]
153.137
153.14
Powder

Chemical Identifiers

2042-14-0
153.137
BQEXDUKMTVYBRK-UHFFFAOYSA-N
16271
CC1=C(C=C(C=C1)O)[N+](=O)[O-]
C7H7NO3
MFCD00007244
3-nitro-4-methylphenol, 3-nitro-p-cresol, 4-methyl-3-nitro-phenol, 4-hydroxy-2-nitrotoluene, 2-nitro-4-hydroxytoluene, phenol, 4-methyl-3-nitro, p-cresol, 3-nitro, unii-4b422hud6h, 4-methyl-5-nitrophenol
4-methyl-3-nitrophenol
Safety and Handling

Safety and Handling

GHS H Statement
H311-H302-H332-H315-H319-H335
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITROCRESOLS, SOLID

EINECSNumber : 218-044-6

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only