missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methyl-3-nitrophenol, 98%

Manufacturer:  Alfa Aesar™ 4428530

 View more versions of this product

Catalog No. AA4428530



Specifications

Specifications

4-Methyl-3-nitrophenol
77°C to 81°C
2042-14-0
MFCD00007244
2-nitro-4-hydroxytoluene
BQEXDUKMTVYBRK-UHFFFAOYSA-N
4-methyl-3-nitrophenol
16271
Powder
Phenol-like
250g
C7H7NO3
UN2446
Slightly soluble in water.
CC1=C(C=C(C=C1)O)[N+](=O)[O-]
153.137
153.14
98%

Chemical Identifiers

2042-14-0
153.137
BQEXDUKMTVYBRK-UHFFFAOYSA-N
16271
CC1=C(C=C(C=C1)O)[N+](=O)[O-]
C7H7NO3
MFCD00007244
2-nitro-4-hydroxytoluene
4-methyl-3-nitrophenol
Safety and Handling

Safety and Handling

GHS H Statement
H311-H302-H332-H315-H319-H335
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITROCRESOLS, SOLID

EINECSNumber : 218-044-6

TSCA : No

Recommended Storage : Ambient temperatures

Documents