missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methyl-3-nitrobenzyl Alcohol, TCI America™

$100.83 - $352.92

Chemical Identifiers

CAS 40870-59-5
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.164
MDL Number MFCD00007177
InChI Key URCWIFSXDARYLY-UHFFFAOYSA-N
PubChem CID 123497
IUPAC Name (4-methyl-3-nitrophenyl)methanol
SMILES CC1=C(C=C(C=C1)CO)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M291625G View Documents TCI America
M291625G
25g Each for $352.92
 
M29165G View Documents TCI America
M29165G
5g Each for $100.83
 
Specifications

Chemical Identifiers

40870-59-5
167.164
URCWIFSXDARYLY-UHFFFAOYSA-N
(4-methyl-3-nitrophenyl)methanol
C8H9NO3
MFCD00007177
123497
CC1=C(C=C(C=C1)CO)[N+](=O)[O-]

Specifications

195°C
43°C
40870-59-5
MFCD00007177
CC1=C(C=C(C=C1)CO)[N+](=O)[O-]
167.164
167.16
≥96.0% (GC)
Yellow
25g
C8H9NO3
URCWIFSXDARYLY-UHFFFAOYSA-N
(4-methyl-3-nitrophenyl)methanol
123497
Crystal-Powder at 20°C
4-Methyl-3-nitrobenzyl Alcohol
Safety and Handling

Safety and Handling

TSCA : No