missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methyl-3-nitrobenzonitrile, TCI America™

$102.36 - $304.03

Chemical Identifiers

CAS 939-79-7
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.148
MDL Number MFCD00031482
InChI Key KOFBNBCOGKLUOM-UHFFFAOYSA-N
Synonym 3-nitro-4-methylbenzonitrile, 3-nitro-p-tolunitrile, 4-methyl-3-nitro-benzonitrile, benzonitrile, 4-methyl-3-nitro, 4-cyano-2-nitrotoluene, 4-methyl-3-nitrobenzenecarbonitrile, 5-cyano-2-methylnitrobenzene, 2-nitro-p-toluonitrile, pubchem4817, acmc-209gtb
PubChem CID 589333
IUPAC Name 4-methyl-3-nitrobenzonitrile
SMILES CC1=C(C=C(C=C1)C#N)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M19465G View Documents TCI America
M19465G
5g Each for $102.36
 
M194625G View Documents TCI America
M194625G
25g Each for $304.03
 
Specifications

Chemical Identifiers

939-79-7
162.148
KOFBNBCOGKLUOM-UHFFFAOYSA-N
589333
CC1=C(C=C(C=C1)C#N)[N+](=O)[O-]
C8H6N2O2
MFCD00031482
3-nitro-4-methylbenzonitrile, 3-nitro-p-tolunitrile, 4-methyl-3-nitro-benzonitrile, benzonitrile, 4-methyl-3-nitro, 4-cyano-2-nitrotoluene, 4-methyl-3-nitrobenzenecarbonitrile, 5-cyano-2-methylnitrobenzene, 2-nitro-p-toluonitrile, pubchem4817, acmc-209gtb
4-methyl-3-nitrobenzonitrile

Specifications

171°C
107°C
939-79-7
MFCD00031482
KOFBNBCOGKLUOM-UHFFFAOYSA-N
4-methyl-3-nitrobenzonitrile
589333
Crystal-Powder at 20°C
4-Methyl-3-nitrobenzonitrile
Undesignated
5g
C8H6N2O2
3-nitro-4-methylbenzonitrile, 3-nitro-p-tolunitrile, 4-methyl-3-nitro-benzonitrile, benzonitrile, 4-methyl-3-nitro, 4-cyano-2-nitrotoluene, 4-methyl-3-nitrobenzenecarbonitrile, 5-cyano-2-methylnitrobenzene, 2-nitro-p-toluonitrile, pubchem4817, acmc-209gtb
CC1=C(C=C(C=C1)C#N)[N+](=O)[O-]
162.148
162.15
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No