missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Methyl-3-nitrobenzonitrile, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC434740050
Specifications
4-Methyl-3-nitrobenzonitrile | |
Brown to White | |
5g | |
96.0 | |
96% min. (HPLC) | |
2-nitro-p-toluonitrile, 3-nitro-4-methylbenzonitrile, 3-nitro-p-tolunitrile, 4-cyano-2-nitrotoluene, 4-methyl-3-nitro-benzonitrile, 4-methyl-3-nitrobenzenecarbonitrile, 5-cyano-2-methylnitrobenzene, acmc-209gtb, benzonitrile, 4-methyl-3-nitro, pubchem4817 | |
CC1=C(C=C(C=C1)C#N)[N+](=O)[O-] | |
162.15 | |
162.15 | |
97% |
Authentic | |
102°C to 106°C | |
939-79-7 | |
100.0 | |
C8H6N2O2 | |
KOFBNBCOGKLUOM-UHFFFAOYSA-N | |
4-methyl-3-nitrobenzonitrile | |
589333 | |
Crystalline Powder |
Chemical Identifiers
939-79-7 | |
162.15 | |
2-nitro-p-toluonitrile, 3-nitro-4-methylbenzonitrile, 3-nitro-p-tolunitrile, 4-cyano-2-nitrotoluene, 4-methyl-3-nitro-benzonitrile, 4-methyl-3-nitrobenzenecarbonitrile, 5-cyano-2-methylnitrobenzene, acmc-209gtb, benzonitrile, 4-methyl-3-nitro, pubchem4817 | |
4-methyl-3-nitrobenzonitrile |
C8H6N2O2 | |
KOFBNBCOGKLUOM-UHFFFAOYSA-N | |
589333 | |
CC1=C(C=C(C=C1)C#N)[N+](=O)[O-] |
Safety and Handling
GHS H Statement
Toxic if inhaled.
Toxic in contact with skin.
Toxic if swallowed.
GHS P Statement
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with ple
GHS Signal Word: Danger