Please login to your online account to display your discounted pricing

4-Methyl-3-nitrobenzonitrile, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Methyl-3-nitrobenzonitrile
CAS 939-79-7
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H6N2O2
Formula Weight 162.15
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC434740050

 
thermo fisher scientific
AC434740050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC434740250

 
thermo fisher scientific
AC434740250
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder
Assay ≥96.0%
Formula Weight 162.15
Quantity 5g
Color White to yellow to pale brown
Melting Point 102° to 108°C
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 4-Methyl-3-nitrobenzonitrile
Infrared Spectrum Authentic
Assay Percent Range 97%
Molecular Formula C8H6N2O2
CAS 939-79-7
Health Hazard 2 GHS H Statement:
Toxic if inhaled.
Toxic in contact with skin.
Toxic if swallowed.
Health Hazard 3 GHS P Statement:
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF exposed or concerned: Get medical advice/attention.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 GHS Signal Word: Danger
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF exposed or concerned: Get medical advice/attention.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if inhaled.
Toxic in contact with skin.
Toxic if swallowed.