Learn More
4-Methyl-3-nitrobenzoic acid, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 165991000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Methyl-3-nitrobenzoic acid | |
96-98-0 | |
100.0 | |
White to Yellow | |
99% | |
C8H7NO4 | |
MFCD00007174 | |
09, 502 | |
Solubility in water: slightly soluble. Other solubilities: soluble in alcohols | |
CC1=C(C=C(C=C1)C(=O)O)[N+](=O)[O-] | |
7319 | |
99% |
99% | |
98.5 | |
187.0°C to 190.0°C | |
Authentic | |
Glass bottle | |
CH3C6H3(NO2)CO2H | |
100 g | |
4-methyl-3-nitrobenzoic acid, 3-nitro-4-methylbenzoic acid, 3-nitro-p-toluic acid, benzoic acid, 4-methyl-3-nitro, 3-nitro-para-toluic acid, m-nitro-p-toluic acid, 4-methyl-3-nitro-benzoic acid, p-toluic acid, 3-nitro, 3-nitro-4-methyl benzoic acid, benzoic acid,4-methyl-3-nitro | |
BBEWSMNRCUXQRF-UHFFFAOYSA-N | |
181.15 | |
181.15 | |
Crystalline Powder |
Chemical Identifiers
96-98-0 | |
181.15 | |
BBEWSMNRCUXQRF-UHFFFAOYSA-N | |
CC1=C(C=C(C=C1)C(=O)O)[N+](=O)[O-] |
C8H7NO4 | |
MFCD00007174 | |
7319 |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
GHS Signal Word: Warning
EINECSNumber : 202-549-3
TSCA : TSCA
RUO – Research Use Only