Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methyl-3-nitrobenzoic acid, 99%

$239.58 - $239.58

Chemical Identifiers

Chemical Name or Material 4-Methyl-3-nitrobenzoic acid
CAS 96-98-0
Molecular Formula C8H7NO4
Formula Weight 181.15
MDL Number MFCD00007174
Beilstein 1874411
Synonym 3-Nitro-p-toluic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1165036

  SDS
alfa aesar™
A1165036
500g Each for $239.58
Description & Specifications

Specifications

Formula Weight 181.15
Chemical Name or Material 4-Methyl-3-nitrobenzoic acid
TSCA TSCA
Assay 99%
Molecular Formula C8H7NO4
MDL Number MFCD00007174
CAS 96-98-0
Beilstein 1874411
Synonym 3-Nitro-p-toluic acid
Health Hazard 1 Warning
Melting Point 188° to 190°C
Quantity 500g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.