Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methyl-3-nitrobenzenesulfonamide, 97%

$118.68 - $118.68

Chemical Identifiers

Chemical Name or Material 4-Methyl-3-nitrobenzenesulfonamide
CAS 6949-23-1
Molecular Formula C7H8N2O4S
Formula Weight 216.22
MDL Number MFCD00460611
Synonym 3-Nitro-p-toluenesulfonamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2603903

 
alfa aesar™
H2603903
1g Each for $118.68
Description & Specifications

Specifications

Formula Weight 216.22
Chemical Name or Material 4-Methyl-3-nitrobenzenesulfonamide
Assay 97%
Molecular Formula C7H8N2O4S
MDL Number MFCD00460611
CAS 6949-23-1
Synonym 3-Nitro-p-toluenesulfonamide
Health Hazard 1 Warning
Melting Point 141° to 144°C
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.