Please login to your online account to display your discounted pricing

4-Methyl-3-nitroanisole, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Methyl-3-nitroanisole
CAS 17484-36-5
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C8H9NO3
Linear Formula CH3C6H3(NO2)OCH3
Formula Weight 167.16
MDL Number MFCD00007173
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC161000050

  SDS
thermo fisher scientific
AC161000050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Assay ≥98.5%
Formula Weight 167.16
Quantity 5g
Color Clear yellow to brown-yellow
Density 1.2g/mL
Beilstein 06, 411
CAS Max % 100.0
Synonym 4-Methoxy-2-nitrotoluene
Chemical Name or Material 4-Methyl-3-nitroanisole
Packaging Glass bottle
Refractive Index 1.5515 to 1.5535 (20°C, 589nm)
Assay Percent Range 99%
Molecular Formula C8H9NO3
Linear Formula CH3C6H3(NO2)OCH3
MDL Number MFCD00007173
CAS 17484-36-5
CAS Min % 98.5
Infrared Spectrum Authentic