Learn More
4-Methyl-3-nitroaniline, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 127251000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
4-Methyl-3-nitroaniline | |
96.0 | |
Authentic | |
Glass bottle | |
CH3C6H3(NO2)NH2 | |
100g | |
4-amino-2-nitrotoluene, 3-nitro-4-methylaniline, 3-nitro-p-toluidine, m-nitro-p-toluidine, 2-nitro-4-aminotoluene, benzenamine, 4-methyl-3-nitro, 5-nitro-4-toluidine, p-toluidine, 3-nitro, gl-amin czech, gl-amin | |
CC1=C(C=C(C=C1)N)[N+](=O)[O-] | |
152.15 | |
CHEBI:81670 | |
97% |
119-32-4 | |
100.0 | |
96% min. (HPLC) | |
C7H8N2O2 | |
MFCD00007910 | |
12, 996 | |
GDIIPKWHAQGCJF-UHFFFAOYSA-N | |
4-methyl-3-nitroaniline | |
8390 | |
152.15 |
Chemical Identifiers
119-32-4 | |
152.15 | |
GDIIPKWHAQGCJF-UHFFFAOYSA-N | |
8390 | |
4-methyl-3-nitroaniline |
C7H8N2O2 | |
MFCD00007910 | |
4-amino-2-nitrotoluene, 3-nitro-4-methylaniline, 3-nitro-p-toluidine, m-nitro-p-toluidine, 2-nitro-4-aminotoluene, benzenamine, 4-methyl-3-nitro, 5-nitro-4-toluidine, p-toluidine, 3-nitro, gl-amin czech, gl-amin | |
CHEBI:81670 | |
CC1=C(C=C(C=C1)N)[N+](=O)[O-] |
Safety and Handling
GHS H Statement:
Toxic to aquatic life with long lasting effects.
May cause damage to organs through prolonged or repeated exposure.
GHS P Statement:
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 204-314-
RUO â Research Use Only