Please login to your online account to display your discounted pricing

4-Methyl-3-heptanol, 99+%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  126890050

 View more versions of this product

Catalog No. AC126890050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.0%
Physical Form Liquid
Formula Weight 130.23
Quantity 5g
Color Clear colorless
EINECS Number 239-058-9
Density 0.8200g/mL
RTECS Number MJ4600000
Beilstein 01, I, 209
CAS Min % 99.0
CAS Max % 100.0
Chemical Name or Material 4-Methyl-3-heptanol
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4290 to 1.4310 (20°C, 589nm)
Name Note 99+%
Assay Percent Range 99+%
Molecular Formula C8H18O
Linear Formula CH3CH2CH2CH(CH3)CH(OH)C2H5
MDL Number MFCD00004568
CAS 14979-39-6
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 4-Methyl-3-heptanol
Name Note 99+%
CAS 14979-39-6
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C8H18O
Linear Formula CH3CH2CH2CH(CH3)CH(OH)C2H5
Formula Weight 130.23
MDL Number MFCD00004568
Accessories

GHS Signal Word: Warning

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Flammable liquid and vapor.

SDS & Safety Documents