Please login to your online account to display your discounted pricing

4-Methyl-3-heptanol, 99+%, ACROS Organics™

$71.54 - $71.54

Chemical Identifiers

Chemical Name or Material 4-Methyl-3-heptanol
Name Note 99+%
CAS 14979-39-6
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C8H18O
Linear Formula CH3CH2CH2CH(CH3)CH(OH)C2H5
Formula Weight 130.23
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC126890050

  SDS
thermo fisher scientific
126890050
Glass bottle 5g Each for $71.54
Description & Specifications

Specifications

Assay ≥99.0%
Formula Weight 130.23
Physical Form Liquid
Quantity 5g
Color Clear colorless
RTECS Number MJ4600000
EINECS Number 239-058-9
Density 0.8200g/mL
Beilstein 01, I, 209
CAS Min % 99.0
CAS Max % 100.0
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Infrared Spectrum Authentic
Refractive Index 1.4290 to 1.4310 (20°C, 589nm)
Health Hazard 2 GHS H Statement:
Flammable liquid and vapor.
Health Hazard 1 GHS Signal Word: Warning
Chemical Name or Material 4-Methyl-3-heptanol
Linear Formula CH3CH2CH2CH(CH3)CH(OH)C2H5
Name Note 99+%
Molecular Formula C8H18O
CAS 14979-39-6
MDL Number MFCD00004568
Assay Percent Range 99+%
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Flammable liquid and vapor.