missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methyl-3-cyclohexene-1-carboxylic Acid, TCI America™

$126.81 - $435.42

Chemical Identifiers

CAS 4342-60-3
Molecular Formula C8H12O2
Molecular Weight (g/mol) 140.182
InChI Key OYOQOLNBTPTFEM-UHFFFAOYSA-N
PubChem CID 20350
IUPAC Name 4-methylcyclohex-3-ene-1-carboxylic acid
SMILES CC1=CCC(CC1)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M24921G View Documents TCI America
M24921G
1g Each for $126.81
 
M24925G View Documents TCI America
M24925G
5g Each for $435.42
 
Specifications

Chemical Identifiers

4342-60-3
140.182
20350
CC1=CCC(CC1)C(=O)O
C8H12O2
OYOQOLNBTPTFEM-UHFFFAOYSA-N
4-methylcyclohex-3-ene-1-carboxylic acid

Specifications

White
1g
C8H12O2
CC1=CCC(CC1)C(=O)O
140.182
140.18
≥98.0% (GC,T)
100°C
4342-60-3
OYOQOLNBTPTFEM-UHFFFAOYSA-N
4-methylcyclohex-3-ene-1-carboxylic acid
20350
Crystal-Powder at 20°C
4-Methyl-3-cyclohexene-1-carboxylic Acid
Safety and Handling

Safety and Handling

TSCA : No