Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methyl-3,5-dinitrobenzeneboronic acid, 97%

$233.55 - $2,315.89

Chemical Identifiers

Chemical Name or Material 4-Methyl-3,5-dinitrobenzeneboronic acid
CAS 28249-49-2
Molecular Formula C7H7BN2O6
Formula Weight 225.95
MDL Number MFCD08060935
Synonym 3,5-Dinitro-4-methylphenylboronic acid; 3,5-Dinitro-4-methylbenzeneboronic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2577303

  SDS
alfa aesar™
H2577303
1g Each for $233.55

AAH2577314

  SDS
alfa aesar™
H2577314
25g Each for $2,315.89
Description & Specifications

Specifications

Formula Weight 225.95
Quantity 1g
Melting Point ≈280°C (dec.)
Synonym 3,5-Dinitro-4-methylphenylboronic acid; 3,5-Dinitro-4-methylbenzeneboronic acid
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 28249-49-2
Assay 97%
Molecular Formula C7H7BN2O6
Chemical Name or Material 4-Methyl-3,5-dinitrobenzeneboronic acid
MDL Number MFCD08060935
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.