Please login to your online account to display your discounted pricing

4-Methyl-3,4-dihydro-2H-1,4-benzoxazine-7-carboxylic acid, 97%, Maybridge

$72.79 - $72.79

Chemical Identifiers

Chemical Name or Material 4-Methyl-3, 4-dihydro-2H-1, 4-benzoxazine-7-carboxylic acid
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H11NO3
Formula Weight 193.2
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC13501DE

 
fisher scientific company
CC13501DE
Amber Glass Bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC13501DA

  SDS
fisher scientific company
CC13501DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC13501CB

  SDS
fisher scientific company
CC13501CB
Amber Glass Bottle 250mg Each for $72.79
Description & Specifications

Specifications

Formula Weight 193.2
Quantity 5g
Assay 97%
Assay Percent Range 97%
Chemical Name or Material 4-Methyl-3, 4-dihydro-2H-1, 4-benzoxazine-7-carboxylic acid
Molecular Formula C10H11NO3
CAS 532391-89-2
Packaging Amber Glass Bottle