Please login to your online account to display your discounted pricing

4-Methyl-3,4-dihydro-2H-1,4-benzoxazine-7-carboxylic acid, 97%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC13501CB

 View more versions of this product

Catalog No. CC13501CB

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 193.2
Quantity 250mg
Assay 97%
Chemical Name or Material 4-Methyl-3, 4-dihydro-2H-1, 4-benzoxazine-7-carboxylic acid
Packaging Amber Glass Bottle
CAS 532391-89-2
Molecular Formula C10H11NO3
Assay Percent Range 97%

Specifications

Chemical Name or Material 4-Methyl-3, 4-dihydro-2H-1, 4-benzoxazine-7-carboxylic acid
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H11NO3
Formula Weight 193.2