missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methyl-3,4-dihydro-2H-1,4-benzoxazine-7-carbaldehyde, 97%, Maybridge

Manufacturer:  Fisher Scientific CC13504DA

 View more versions of this product

Catalog No. CC13504DA



Specifications

Specifications

4-Methyl-3, 4-dihydro-2H-1, 4-benzoxazine-7-carbaldehyde
250mg
96.5
97%
MFCD02681912
WJHBKMPLXRGBGM-UHFFFAOYSA-N
4-methyl-2,3-dihydro-1,4-benzoxazine-7-carbaldehyde
2776404
97%
Amber Glass Bottle
141103-93-7
100.0
C10H11NO2
4-methyl-3,4-dihydro-2h-1,4-benzoxazine-7-carbaldehyde, 4-methyl-3,4-dihydro-2h-benzo b 1,4 oxazine-7-carbaldehyde, 2h-1,4-benzoxazine-7-carboxaldehyde,3,4-dihydro-4-methyl, 2h-1,4-benzoxazine-7-carboxaldehyde, 3,4-dihydro-4-methyl, acmc-20n02i, 3,4-dihydro-7-formyl-4-methyl-2h-1,4-benzoxazine, 3,4-dihydro-4-methyl-2h-1,4-benzoxazine-7-carboxaldehyde, 4-methyl-2,3-dihydro-2h-1,4-benzoxazine-7-carbaldehyde, 4-methyl-3,4-dihydro-2h-benzo 1,4 oxazine-7-carbaldehyde, 4-methyl-2h,3h-benzo e 1,4-oxazaperhydroine-7-carbaldehyde
CN1CCOC2=C1C=CC(=C2)C=O
177.203
177.2

Chemical Identifiers

141103-93-7
177.203
WJHBKMPLXRGBGM-UHFFFAOYSA-N
2776404
CN1CCOC2=C1C=CC(=C2)C=O
C10H11NO2
MFCD02681912
4-methyl-3,4-dihydro-2h-1,4-benzoxazine-7-carbaldehyde, 4-methyl-3,4-dihydro-2h-benzo b 1,4 oxazine-7-carbaldehyde, 2h-1,4-benzoxazine-7-carboxaldehyde,3,4-dihydro-4-methyl, 2h-1,4-benzoxazine-7-carboxaldehyde, 3,4-dihydro-4-methyl, acmc-20n02i, 3,4-dihydro-7-formyl-4-methyl-2h-1,4-benzoxazine, 3,4-dihydro-4-methyl-2h-1,4-benzoxazine-7-carboxaldehyde, 4-methyl-2,3-dihydro-2h-1,4-benzoxazine-7-carbaldehyde, 4-methyl-3,4-dihydro-2h-benzo 1,4 oxazine-7-carbaldehyde, 4-methyl-2h,3h-benzo e 1,4-oxazaperhydroine-7-carbaldehyde
4-methyl-2,3-dihydro-1,4-benzoxazine-7-carbaldehyde
SDS Documents