missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methyl-3,4-dihydro-2H-1,4-benzoxazine-7-carbaldehyde, 97%, Maybridge

$191.92 - $191.92

Chemical Identifiers

CAS 141103-93-7
Molecular Formula C10H11NO2
Molecular Weight (g/mol) 177.203
MDL Number MFCD02681912
InChI Key WJHBKMPLXRGBGM-UHFFFAOYSA-N
Synonym 4-methyl-3,4-dihydro-2h-1,4-benzoxazine-7-carbaldehyde, 4-methyl-3,4-dihydro-2h-benzo b 1,4 oxazine-7-carbaldehyde, 2h-1,4-benzoxazine-7-carboxaldehyde,3,4-dihydro-4-methyl, 2h-1,4-benzoxazine-7-carboxaldehyde, 3,4-dihydro-4-methyl, acmc-20n02i, 3,4-dihydro-7-formyl-4-methyl-2h-1,4-benzoxazine, 3,4-dihydro-4-methyl-2h-1,4-benzoxazine-7-carboxaldehyde, 4-methyl-2,3-dihydro-2h-1,4-benzoxazine-7-carbaldehyde, 4-methyl-3,4-dihydro-2h-benzo 1,4 oxazine-7-carbaldehyde, 4-methyl-2h,3h-benzo e 1,4-oxazaperhydroine-7-carbaldehyde
PubChem CID 2776404
IUPAC Name 4-methyl-2,3-dihydro-1,4-benzoxazine-7-carbaldehyde
SMILES CN1CCOC2=C1C=CC(=C2)C=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC13504DA SDS View Documents Fisher Scientific Company
CC13504DA
250mg Amber Glass Bottle Each for $191.92
 
Specifications

Chemical Identifiers

141103-93-7
177.203
WJHBKMPLXRGBGM-UHFFFAOYSA-N
2776404
CN1CCOC2=C1C=CC(=C2)C=O
C10H11NO2
MFCD02681912
4-methyl-3,4-dihydro-2h-1,4-benzoxazine-7-carbaldehyde, 4-methyl-3,4-dihydro-2h-benzo b 1,4 oxazine-7-carbaldehyde, 2h-1,4-benzoxazine-7-carboxaldehyde,3,4-dihydro-4-methyl, 2h-1,4-benzoxazine-7-carboxaldehyde, 3,4-dihydro-4-methyl, acmc-20n02i, 3,4-dihydro-7-formyl-4-methyl-2h-1,4-benzoxazine, 3,4-dihydro-4-methyl-2h-1,4-benzoxazine-7-carboxaldehyde, 4-methyl-2,3-dihydro-2h-1,4-benzoxazine-7-carbaldehyde, 4-methyl-3,4-dihydro-2h-benzo 1,4 oxazine-7-carbaldehyde, 4-methyl-2h,3h-benzo e 1,4-oxazaperhydroine-7-carbaldehyde
4-methyl-2,3-dihydro-1,4-benzoxazine-7-carbaldehyde

Specifications

Amber Glass Bottle
141103-93-7
100.0
C10H11NO2
4-methyl-3,4-dihydro-2h-1,4-benzoxazine-7-carbaldehyde, 4-methyl-3,4-dihydro-2h-benzo b 1,4 oxazine-7-carbaldehyde, 2h-1,4-benzoxazine-7-carboxaldehyde,3,4-dihydro-4-methyl, 2h-1,4-benzoxazine-7-carboxaldehyde, 3,4-dihydro-4-methyl, acmc-20n02i, 3,4-dihydro-7-formyl-4-methyl-2h-1,4-benzoxazine, 3,4-dihydro-4-methyl-2h-1,4-benzoxazine-7-carboxaldehyde, 4-methyl-2,3-dihydro-2h-1,4-benzoxazine-7-carbaldehyde, 4-methyl-3,4-dihydro-2h-benzo 1,4 oxazine-7-carbaldehyde, 4-methyl-2h,3h-benzo e 1,4-oxazaperhydroine-7-carbaldehyde
CN1CCOC2=C1C=CC(=C2)C=O
177.203
177.2
4-Methyl-3, 4-dihydro-2H-1, 4-benzoxazine-7-carbaldehyde
250mg
96.5
97%
MFCD02681912
WJHBKMPLXRGBGM-UHFFFAOYSA-N
4-methyl-2,3-dihydro-1,4-benzoxazine-7-carbaldehyde
2776404
97%