Please login to your online account to display your discounted pricing

4-Methyl-3,4-dihydro-2H-1,4-benzoxazine-6-sulfonyl chloride, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 4-Methyl-3, 4-dihydro-2H-1, 4-benzoxazine-6-sulfonyl chloride
CAS 892948-94-6
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H10ClNO3S
Formula Weight 247.7
MDL Number MFCD10700049
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC36103DA

  SDS
fisher scientific company
CC36103DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC36103CB

  SDS
fisher scientific company
CC36103CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 247.7
Quantity 1g
Assay 97%
CAS Max % 100.0
Chemical Name or Material 4-Methyl-3, 4-dihydro-2H-1, 4-benzoxazine-6-sulfonyl chloride
Assay Percent Range 97%
CAS 892948-94-6
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
Molecular Formula C9H10ClNO3S
MDL Number MFCD10700049
CAS Min % 96.5
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Amber Glass Bottle
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.